Crystal violet BS

Supplier: Ambeed
Danger

Synonyms: Basic Violet 3GentiavermMethylrosanilide chlorideHexamethyl VioletVianinGentian violetMethyl violet 10BCI 42555Hexamethyl-p-rosaniline chlorideN-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chlorideDimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-yliden)ammoniumchloridViocid

A1003341-25G A1003341-500G
77595-648EA 74 USD
77595-648 76646-116
Crystal violet BS
Crystal Violet

Caution: for Research Use Only

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Number: MFCD00011750
CAS Number: 548-62-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR