Periodic acid for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Orthoperiodic acidortho-Periodic acid

453170250 453175000 45317-1000
TS45317-1000EA 148.45 USD
TS45317-1000 TS453170250 TS453175000
Periodic acid for analysis
Periodic acid
Formula: H₅IO₆
MW: 227.94 g/mol
Melting Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00011344
CAS Number: 10450-60-9
UN: 3085
ADR: 5.1,II
https://us.vwr.com/stibo/web/std.lang.all/90/77/779077.jpg

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Titration Iodimetric 99.0 to 101.0 %
Residue after ignition ≤0.05 %
Iodide (I) ≤10 ppm
Bromide and Bromate (as Cl) ≤200 ppm
Chloride and Chlorate (as Cl) ≤200 ppm
Nitrate (NO3) ≤100 ppm
Sulfate (SO4) ≤100 ppm
Calcium (Ca) ≤50 ppm
Copper (Cu) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Nickel (Ni) ≤50 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤100 ppm
Zinc (Zn) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm
Insoluble matter ≤0.01 %
Iodic acid ≤1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR