Hydrobromic acid 33% (w/w) in glacial acetic acid, AcroSeal™, pure

Supplier: Thermo Scientific Chemicals
Danger

43069-1000 430698000
TS43069-1000EA 52 USD
TS43069-1000 TS430698000
Hydrobromic acid 33% (w/w) in glacial acetic acid, AcroSeal™, pure
Hydrogen bromide
Formula: HBr
Density: 1.42 g/cm³
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to yellow-orange Solution (May darken on storage)
Water ≤0.5 %
Titration with NaOH 31.5 to 36.5 %
HPLC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR