Potassium-tert-butoxide 1 M in tert-butanol, AcroSeal™

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium tert-butanolatePotassium t-butoxidePotassium tert-butoxidePotassium tert-butylate

42612-1000
TS42612-1000EA 90.1 USD
TS42612-1000
Potassium-tert-butoxide 1 M in tert-butanol, AcroSeal™
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
MDL Number: MFCD00012162
CAS Number: 865-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR