Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

42395-0025 423950010 423950250 423955000
TS42395-0025EA 99 USD
TS42395-0025 TS423950010 TS423950250 TS423955000
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Methanol
Formula: CH₃OH
Boiling Pt: 64.7 °C (760 mmHg)
Melting Pt: –98 °C
Density: 0.791 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 % (K.F.)
Boiling point 64 to 65 °C
Density (20/4 °C) 0.791 to 0.793
Solubility (In water) Passes test
GC ≥99.8 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Carbonyl compounds Each of acetone, formaldehyde and acetaldehyde
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0002 meq/g
KMnO4-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR