Petrolatum, white, pure
Supplier: Thermo Scientific Chemicals
41709-0010
417090100
TS41709-0010EA
51
USD
TS41709-0010
TS417090100
Petrolatum, white, pure
Petrolatum
Boiling Pt:
300 °C Melting Pt: 50…56 °C Density: 0.84 g/cm³ Flash Pt: >150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00147839 CAS Number: 8009-03-8 |
Specification Test Results
Appearance | White Paste |
Infrared spectrum | Conforms |
Viscosity | 5 to 8 mm²/s (at 100 °C) |
Solidification point | 50 to 56°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...