Chloroform 99+% stabilised for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Stabilised with ethanol

39076-0010 390760025
TS39076-0010EA 98 USD
TS39076-0010 TS390760025
Chloroform 99+% stabilised for HPLC
Chloroform
Formula: CHCl₃
Boiling Pt: 61 °C (760 mmHg)
Melting Pt: –63 °C
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM
New Product

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.01 % (K.F.)
GC ≥99.0 %
UV at 245 nm A: ≤1.00
Color scale ≤10 APHA
Stabilizer 0.5 to 1.0 % Ethanol
Residue after evaporation ≤0.0002 %
Free chlorine ≤0.00003 % (as Cl)
Free acid (HCl) ≤0.001 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR