Orthophosphoric acid 98+%, pure

Supplier: Thermo Scientific Chemicals
Danger

38902-1000 389020025 389025000
TS38902-1000EA 63 USD
TS38902-1000 TS389020025 TS389025000
Orthophosphoric acid 98+%, pure
Orthophosphoric acid
Formula: H₃PO₄
Boiling Pt: ∼158 °C (1013 hPa)
Melting Pt: 42 °C
Density: 1.7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 3453
ADR: 8,III

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration with NaOH ≥98.0 %
Sulfate (SO4) ≤30 ppm
Iron (Fe) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR