n-Hexane, dry_extra over Molecular Sieve 97%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

36437-0010 36437-1000 364370025 364375000
TS36437-0010EA 120 USD
TS36437-0010 TS36437-1000 TS364370025 TS364375000
n-Hexane, dry_extra over Molecular Sieve 97%, AcroSeal™
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.005 % (Coulometric)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR