Acetonitrile 99.9% for residue analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

32675-0010 326750025
TS32675-0010EA 121 USD
TS32675-0010 TS326750025
Acetonitrile 99.9% for residue analysis
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –46 °C
Density: 0.781 g/cm³
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
GC ≥99.9 %
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity (CH3COOH) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR