Methanol 99.9%, gradient grade for HPLC

Supplier: Thermo Scientific Chemicals
Danger

32574-0010 325740025
TS32574-0010EA 85 USD
TS32574-0010 TS325740025
Methanol 99.9%, gradient grade for HPLC
Methanol
Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.791 g/cm³
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 %
Additional info filtered through 0.2 u membrane filter
GC ≥99.9 %
UV 1 cm cell vs HPLC-grade water
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g
Gradient analysis :
Absorbance largest eluted peak ≤0.001 A.U.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR