Polyethylene glycol 1,500
Supplier: Thermo Scientific Chemicals
19226-0010
192260051
192261000
TS19226-0010EA
84
USD
TS19226-0010
TS192260051
TS192261000
Polyethylene glycol 1,500
Polyethylene glycol 1,500
Formula:
H(OCH₂CH₂)nOH Melting Pt: 44…48 °C Flash Pt: >250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Specification Test Results
Appearance | White Waxy flakes |
Infrared spectrum | Conforms |
Water | ≤1.0 % (K.F.) |
Hydroxyl value | 70 to 80 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...