m-Xylene, extra dry 99%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
15893-1000
158930010
TS15893-1000EA
51
USD
TS15893-1000
TS158930010
m-Xylene, extra dry 99%, AcroSeal™
m-Xylene
Formula:
C₈H₁₀ Boiling Pt: 139 °C (760 mmHg) Melting Pt: –48 °C Density: 0.864 g/cm³ Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008536 CAS Number: 108-38-3 UN: 1307 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.005 % |
GC | ≥98.5 % |
Residue after evaporation | ≤0.01 % |
Heavy metals (as Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...