Oxalic acid solution_standard 0.1 N
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethanedioic acid
12428-0010
124280025
TS12428-0010EA
54
USD
TS12428-0010
TS124280025
Oxalic acid solution_standard 0.1 N
Oxalic acid
Formula:
C₂H₂O₄ MW: 90.04 g/mol Boiling Pt: 365 Density: 1 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00002573 CAS Number: 144-62-7 EINECS: 205-634-3 Merck Index: 13,06980 |
Specification Test Results
Appearance | Clear colorless Solution |
Strength | 0.0998 to 0.1002 N (20 °C) |
Melting point | 116 to 119 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...