Microscope Slide Storage Cabinets, Four E's USA
Supplier: FOUR E#S INDUSTRIAL GROUP INC.
SSC200204 SSC100104
76644-970EA 144.12 USD
76644-970 76644-972 76644-950 76644-952 76644-954 76644-956 76644-958 76644-960 76644-962 76644-964 76644-966 76644-968
Microscope Slide Storage Cabinets, Four E's USA
Slide Holders
Four E's is a leading manufacturer of microscope slide storage cabinets.
  • Store up to 5000 microscope slides per cabinet
  • Durable cold rolled steel construction with epoxy paint
  • 10 year warranty
These slide storage cabinets are designed for long term storage of large quantities of microscope slides in histology laboratories and hospitals. These cabinets have 6 drawers that hold up to 5000 standard 1×3" microscope slides. These cabinets come with interlocking system for safe stacking and can be stacked safely up to 12 units high. They are made of cold rolled steel with epoxy powder coating. The drawers are designed to slide in and out smoothly. Each drawer comes with two foam blocks to maintain slides upright when drawers are partially filled. Base without legs available. Cabinet and base available in Tan.
Ordering information: Cabinets and bases are sold separately. Ten-year warranty.
Delivery information: Includes foam drawer inserts.
Packaging: Packaged in sturdy cardboard box.