Welch Allyn® 767 Mobile Aneroid Sphygmomanometer
Supplier: Healthcare Products
WEL 7670-03 WEL 7670-06P
470347-186EA 903.12 USD
470347-186 470347-304
Welch Allyn® 767 Mobile Aneroid Sphygmomanometer
Blood Pressure Monitors
Wall or stand use.
  • Built-in cuff storage basket
  • Laser-engraved dial face
  • Recessed dial face and high contrast red pointer increase visibility and reduce parallax errors
  • Five-leg stand provides improved stability and maneuverability
  • Lifetime calibration warranty
  • Mobile aneroid sphygmomanometer with one piece adult cuff
Based on Welch Allyn’s near century of experience building and designing innovative blood pressure technology, the versatile 767 Aneroid is the smart choice for accurate and reliable readings throughout your facility. Improved ergonomicsMore Product Information