Baxter® ELI 380 Resting ECG
Supplier: Healthcare Products
WELELI380-ACX11 WEL ELI380-DCS11
470347-252EA 35432.48 USD
470347-252 470347-258
Baxter® ELI 380 Resting ECG
Data Loggers
Reduce the need for repeat ECGs.
  • Streamlined barcode workflow eliminates steps and reduces manual entry
  • Glass surfaces on the capacitive touchscreen display simplify cleaning
  • Follows ACC, AHA and HRS recommendations for baseline filtering (0.05 Hz)
  • Exceeds ACC, AHA and HRS upper frequency cutoff (300 Hz) for adults and children
  • DoD-approved security features safeguard patient health information (PHI) and your network
  • Critical Test Results help quickly identify high-risk patients
  • 12-lead or 15-lead ECG
The ELI380 Resting ECG, with ERGO swivel and touchscreen display, is designed to make the total ECG experience more efficient-from decreasing the time required to perform the procedure, to increasing the accuracy and security of the data acquired, to the overall patient care experience.