Life/form® Cricothyrotomy Simulator
Supplier: Nasco
LF00994 LF00994A LF00994B LF00994C
470348-710EA 1201.8 USD
470348-710 470348-712 470348-666 470348-716
Life/form® Cricothyrotomy Simulator
Models Human Biology Models
Adult and child practice.
  • Includes carry case
  • 5-year warranty
The Life/form® cricothyrotomy simulator is developed for learning and practicing the techniques necessary to perform needle or surgical cricothyrotomy procedures. The simulator is used by paramedics, EMTs, combat medics, flight nurses, anesthesiologists, and other emergency medical personnel to strengthen the ability and confidence to perform or assist in implementing surgical airways. Anatomically accurate landmarks aid in site training and allow for fast action.
The hyperextended neck allows the user to determine the proper incision site. The trachea in the simulator is replaceable, as the airway passes completely through from top to bottom. This allows checking the stylet and obturator placement once the incision has been made. Complete with a chin and full-size neck, ties which can be used to hold the obturator in a secure position and inflation of the simulated lung verifies correct placement.