Spot Vital Signs 4400, Welch Allyn®
Supplier: Healthcare Products
Welch Allyn®
WEL44WT-B WEL77790-4 WEL4400-PS WEL44XT-B WEL4400-MBS WEL4400-DST
470347-264EA 4640.05 USD
470347-264 470347-088 470347-074 470347-076 470347-136 470347-116
Spot Vital Signs 4400, Welch Allyn®
Clinical Diagnostic Supplies Clinical Instruments
Save time and help reduce data entry errors.
  • Record blood pressure in just 15 seconds
  • SureTemp Plus oral/axillary thermometry
  • 7" color touchscreen
  • EMR connectivity to avoid documentation errors
You work in a fast-paced environment, so you can’t afford to slow down when it comes to capturing the basics. Our next-generation vital signs device is here to help. Experience simple, accurate vitals capture with a solution that fits smoothly into your daily routine. The Spot Vital Signs 4400 device offers an efficient way to capture, access and document patient vitals so you can focus on what really matters—spending more time with your patients.