X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)
Supplier: MilliporeSigma
Synonyms:
X-Gluc
203783-100MG
203783-100MG
EM203783-100MGEA
508.37
USD
CA80050-480
EM203783-100MG
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)
X-GlcA ((5-Bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)
Formula:
C₂₀H₂₆BrClN₂O₇ MW: 521.79 g/mol |
MDL Number:
MFCD00058543 CAS Number: 114162-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...