L(+)-Glutamic acid pH 3.0 - 3.5 (8.6 g/l, H₂O, 25 °C) for biochemistry
Supplier: MilliporeSigma
Synonyms:
L-Glutamic acid
, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid
, L(+)-α-Glutamic acid
, H-Glu-OH
, (S)-(+)-Glutamic acid
, (S)-(+)-α-Glutamic acid
, L(+)-1-Aminopropane-1,3-dicarboxylic acid
, (S)-(+)-2-Aminopentanedioic acid
, L(+)-2-Aminopentanedioic acid
1.00291.0250
EM1.00291.0250EA
160.12
USD
EM1.00291.0250
L(+)-Glutamic acid pH 3.0 - 3.5 (8.6 g/l, H₂O, 25 °C) for biochemistry
L(+)-Glutamic acid
Formula:
HO₂CCH₂CH₂CH(NH₂)CO₂H MW: 147.13 g/mol |
MDL Number:
MFCD00002634 CAS Number: 56-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...