Heptane (mixture of isomers) ≥99% (n-heptane), Supelco®
Supplier: MilliporeSigma
HX0080A-20
EM-HX0080A-20EA
12253.91
USD
EM-HX0080A-20
Heptane (mixture of isomers) ≥99% (n-heptane), Supelco®
Heptane (mixture of isomers)
Formula:
H₃C(CH₂)₅CH₃ MW: 100.2 g/mol |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 |
Specification Test Results
Assay | ≥99% n-heptane basis |
Assay | ≥99.9% (saturated C7 hydrocarbons) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...