Potassium chloride, Molecular Biology Grade
Supplier: MilliporeSigma
529552-250GM
529552-1KG
80503-244EA
84.43
USD
80503-244
80503-242
Potassium chloride, Molecular Biology Grade
Potassium chloride
Formula:
KCl |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...