L(-)-Tryptophan

Supplier: MilliporeSigma

Synonyms: L-α-Amino-3-indolepropionic acid(S)-2-Amino-3-(3-indolyl)propionic acidH-Trp-OHL-TryptophanTryptophan

6540-100GM 6540-100GM
80058-790EA 196.14 USD
CA80058-790 80058-790
L(-)-Tryptophan
L(-)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
MDL Number: MFCD00064340
CAS Number: 73-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR