D-(+)-Trehalose dihydrate
Supplier: MilliporeSigma
Synonyms:
1-O-α-D-Glucopyranosyl-α-D-glucopyranoside
, Mycose
, α-D-Glucopyranosyl-α-D-glucopyranoside
, α,α-Trehalose
, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
625625-50GM
625625-50GM
80058-758EA
110.7
USD
CA80058-758
80058-758
D-(+)-Trehalose dihydrate
D(+)-Trehalose dihydrate
Formula:
C₁₂H₂₂O₁₁·2H₂O MW: 378.33 g/mol |
MDL Number:
MFCD00071594 CAS Number: 6138-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...