Taurocholic acid sodium salt

Supplier: MilliporeSigma

Synonyms: Cholaic acid sodium saltSodium taurocholateSodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulfonate (1:1)Taurocholic acid sodiumSodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate (1:1)

580217-5GM 580217-25GM 580217-25GM
80058-702EA 660.21 USD
80058-702 80058-704 CA80058-702
Taurocholic acid sodium salt
Taurocholic acid sodium salt
Formula: C₂₆H₄₄NNaO₇S
MW: 537.688 g/mol
MDL Number: MFCD00013050
CAS Number: 145-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR