Cholesterol, highly purified

Supplier: MilliporeSigma

Synonyms: CholesterinCholest-5-en-3-ol(3Beta)-Cholest-5-en-3-ol

228111-5GM
80057-932EA 107.5 USD
80057-932
Cholesterol, highly purified
Cholesterol
Formula: C₂₇H₄₆O
MW: 386.66 g/mol
MDL Number: MFCD00003646
CAS Number: 57-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR