ALCH SDA 40B 200 Proof (denatured with Denatonium benzoate)
Supplier: TILLEY CHEMICAL CO., INC
|
Danger
|
Synonyms:
Reagent alcohol
DNA045
DNA047
DNA048
TLDNA045EA
117878.94
USD
TLDNA045
TLDNA047
TLDNA048
ALCH SDA 40B 200 Proof (denatured with Denatonium benzoate)
Alcohol
Storage Temperature:
Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 UN: 1170 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...