Ethanol 190 Proof USP, pure
Supplier: TILLEY CHEMICAL CO., INC
|
Danger
|
Synonyms:
Ethyl alcohol
DNA013
DNA013TP
DNA020
DNA020TP
DNA074
DNA074TP
TLDNA013EA
115247.38
USD
TLDNA013
TLDNA013TP
TLDNA020
TLDNA020TP
TLDNA074
TLDNA074TP
Ethanol 190 Proof USP, pure
Ethanol
Formula:
C₂H₅OH Storage Temperature: Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 UN: 1170 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...