HYDRANAL™ - NEXTGEN Coulomat AG-FI, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Supplier: Honeywell Research Chemicals
34433-500ML 34433-6X500ML
BJ34433-500MLEA 155 USD
BJ34433-500ML BJ344336X500ML
HYDRANAL™ - NEXTGEN Coulomat AG-FI, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Karl Fischer Titrants and Reagents
Anolyte for coulometric Karl Fischer titration, for cells with and without diaphragm, Methanol-based, free of Imidazole (free of CMR substances).
  • Imidazole-free
  • Assay of water: 5 - 50 ppm
  • Methanol as solvent
The suffix “FI”, added to the name of the reagent, indicates that the reagent is “free of imidazole”.
Instead of imidazole, an alternative base has been used which does not compromise the performance of the Hydranal reagent.