PCR Plate Lids, Auto-Sealing, Azenta Life Sciences
Supplier: Azenta Life Sciences
4TI-0291
76638-176CS 220.43 USD
76638-176
PCR Plate Lids, Auto-Sealing, Azenta Life Sciences
Lids Microplate Lids
Low profile, with integrated compression pad, white, no cut corner, for PCR applications on integrated instruments. Developed to support fully automated sealing in the absence of specific instrumentation. Suitable for automation and low throughput workflows.
  • Rigid polycarbonate frame padded with an elastic foam
  • No cut corners
  • Stackable
  • Universal fit to PCR plates
  • Recommended when full sealing automation is required in the absence of dedicated instrumentation
  • Alternative to heat sealing, when heat sealing materialsand instrumentation are not an option
  • Alternative to standard lids for longer incubations
Meets the needs of customers using integrated platforms that often lack suitable heat sealing instrumentation especially when low throughput is needed.
Specialized alternative to standard lids: while standard lids protect reagents, the auto-sealing PCR lid helps minimize reagent evaporation during longer incubations.
Universal fit: due to the lack of cut corners, the lid can be applied to most plates, showing good sealing results when pushed down onto the plate.