Cell Culture Adhesive Seals, Gas Permeable, Peelable

Supplier: AZENTA US, INC
4TI-0517 4TI-0517/ST
76637-908CS 152.8 USD
76637-908 76637-932
Cell Culture Adhesive Seals, Gas Permeable, Peelable
Seals Microplate Seals
The cell culture adhesive seal is a gas permeable adhesive seal which seals assay and tissue culture plates, microplates and storage plates.

  • Peelable
  • Gas permeability rate: 8900 m³/m²/day
  • Moisture vapor transmission rate (very low): 4200 g/m²/day
  • Air porosity: 10 sec/100 cc/in²
  • Due to its paper-based material it should not be used in wet conditions
  • Suitable for cell culture and enables long term culture with significantly reduced evaporation
  • Temperature range: –20 to +40 °C
  • Non sterile

Adhesive seals provide a versatile, yet robust, method for protecting samples.

To obtain the best sealing results with adhesive seals, we strongly recommend the use of high quality plates with raised rim sealing rings for optimal sealing integrity and guaranteed flatness. All Azenta PCR plates are designed with these features.

Accessories information: Adhesive seal roller; ensures consistent seal application across all wells, 1 roller.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR