Clear Heat Sealing Films, Pierceable, Peelable, Gas Permeable
Supplier: AZENTA US, INC
4TI-0540/SLIT 4TI-0540/SLIT/S 4TI-0541/SLIT
76637-786EA 1799.64 USD
76637-786 76637-906 76637-810
Clear Heat Sealing Films, Pierceable, Peelable, Gas Permeable
Seals Microplate Seals
The clear heat seal, peelable film is based on our clear heat seal, with the addition of 3 mm slits across the width of the seal making the seal gas permeable. Suitable for insect and seed storage.
  • Compatible with polypropylene, polystyrene and cyclic olefin copolymer (COC) plates
  • Pierceable with a pipette tip, manually or by liquid handling robots
  • Temperature range: –80 to +110 °C
  • Non sterile
Heat sealing offers a 100% effective method of plate sealing, for complete seal integrity, as well as being quick and cost effective.
The sheet seals fit allMore Product Information