Heat Sealing Films Plus, Clear, High Tensile Strength
Supplier: AZENTA US, INC
4TI-0548 4TI-0548/S 4TI-05481 4TI-0549 4TI-0549/S
76637-952EA 1012.09 USD
76637-952 76637-838 76637-794 76637-914 76637-796
Heat Sealing Films Plus, Clear, High Tensile Strength
Seals Microplate Seals
The clear heat seal plus is a high tensile strength, optically clear polymer film forms a peelable seal to polypropylene, polyethylene, polystyrene, and cyclic olefin copolymer (COC) SBS format plates.
  • Suitable for bead mill applications
  • Forms a complete seal to a plate also enabling short term sample storage
  • Thicker than standard clear heat seal for application requiring high tensile strength, but not as suitable for optical applications due to decreased transmission through the thicker sealing layers
  • Temperature range: –80 to +80 °C
  • Non sterile
Heat sealing offers a 100% effective method for plate sealing for a complete seal integrity, as well as being quick and cost effective. The clear heat seals plus has high tensile strength for sealing of microplates during homogenization or disruption of seeds or other material such as bead mill applications.
The sheet seals fit all standard SBS footprint PCR and qPCR plates, microplates, assay and storage plates, from 12 to 1536 wells.
Ordering information: Available as sheets compatible with manual and semiautomated sealers. Available in multiple roll formats compatible with specified automated heat sealers. Gamma treatment available upon request.