Anti-CD5 Rabbit Monoclonal Antibody [clone: SP19]
Supplier: Diagnostic BioSystems
RMAB011 RMAB011-05-RUO RMAB011-01 RMAB011-RUO RMAB011-05 RMAB011-01-RUO
76634-736EA 1434.32 USD
76634-736 CA76634-734 CA76634-736 76634-732 76634-734 CA76634-732
Anti-CD5 Rabbit Monoclonal Antibody [clone: SP19]
Antibodies
This antibody recognizes CD5, which is a transmembrane protein, is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5(+) while the follicle center cell lymphoma are CD5(-).