Anti-CD20 B-cell Mouse Monoclonal Antibody [clone: L26]
Supplier: Diagnostic BioSystems
PDM004-RUO PDM004
76634-260EA 269.92 USD
CA76634-260 76634-260
Anti-CD20 B-cell Mouse Monoclonal Antibody [clone: L26]
Antibodies
This antibody is specific to a 33 kDa polypeptide present on the majority of B cells in peripheral blood and lymphoid tissue. No reactivity with other hematopoietic cells has been observed.