Polyethylene glycol 8,000 for molecular biology

Supplier: RPI

Synonyms: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000

P48080-1000.0 P48080-10000.0 P48080-25000.0 P48080-500.0 P48080-5000.0 P48080-15000.0
76347-842EA 57.97 USD
76347-842 76347-844 76347-846 76347-848 76347-850 77510-404
Polyethylene glycol 8,000 for molecular biology
Polyethylene glycol 8,000

Ultrapure white crystals for molecular biology applications such as precipitation of DNA, nucleic acids, hybridization and fusion of mammalian cells.


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: H(OCH₂CH₂)nOH
Melting Pt: 55°…60 °C
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Appearance White waxy powder
Color, PTCO ≤ 18
Molecular Weight 7000 - 9000 g/mol
pH (5% in water) 4.5 - 7.5
Viscosity 500 - 900 Centi Stokes
Water ≤ 1.00 %
BSE/TSE Free Conform


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR