Anti-Ezrin/p181/Cytovillin Mouse Monoclonal Antibody [clone: 3C12]
Supplier: Diagnostic Biosystems
MOB380-05 MOB380
76633-786EA 519.63 USD
76633-786 76633-788 76633-784
Anti-Ezrin/p181/Cytovillin Mouse Monoclonal Antibody [clone: 3C12]
Antibodies
This antibody reacts with a protein of 81 kDa. Ezrin serves as a tyrosine kinase substrate and is phosphorylated in EGF-stimulated cells. Ezrin is a cytoplasmic protein, enriched in microvilli and other cell surfaces. Ezrin has an actin-binding capacity. Ezrin is expressed in epithelial cells but not in mesenchymal cells. Ezrin is also expressed by certain epithelial tumors, such as renal cell adenocarcinomas.