Anti-Mast Cell Chymase Mouse Monoclonal Antibody [clone: CC1]
Supplier: Diagnostic Biosystems
MOB346-01 MOB346-05 MOB346
76633-722EA 121 USD
76633-722 76633-724 76633-726
Anti-Mast Cell Chymase Mouse Monoclonal Antibody [clone: CC1]
Antibodies
This antibody reacts with mast cells distributed in skin, synovium, lung, and heart. Mast cells contain a number of preformed chemicals mediators such as histamine, chymase, carboxypeptidase, and proteolytic tryptase. Human mast cell chymase is considered to be an important marker of mast cells and is an important mediator of inflammation.