Anti-Factor XIIIa Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP103 RP103-01 RP103-RUO RP103-05 RP103-01-RUO RP103-05-RUO
76635-120EA 105.1 USD
76635-120 76635-124 CA76635-122 76635-122 CA76635-120 CA76635-124
Anti-Factor XIIIa Rabbit Polyclonal Antibody
Antibodies
This antibody is specific to 160 kDa protein known as Factor XIII-A. Factor XIII is a β-globulin found in plasma and is composed of two subunits, Factor XIII-A and Factor XIII-B. Factor XIII-A is the catalytic subunit and is a dimer of M.W. 160 kDa. Factor XIII is a dermal dendrocyte marker and v variable reaction with these types of tumors.