Anti-ERG Rabbit Monoclonal Antibody [clone: EP111]
Supplier: Diagnostic Biosystems
RMPD034 RMPD034
10065-052EA 297.96 USD
10065-052 76634-864
Anti-ERG Rabbit Monoclonal Antibody [clone: EP111]
Antibodies
ERG, the ETS related gene, belongs to the ETS family that plays important roles in cell development, differentiation, proliferation, apoptosis and tissue remodeling. The ERG antibody identifies endothelial cells, lymphocytes and prostate cancer cells.