Anti-CD10 CALLA Mouse Monoclonal Antibody [clone: 56C6]
Supplier: Diagnostic BioSystems
MOB240-05-RUO MOB240 MOB240-RUO MOB240-05 MOB240-01 MOB240-01-RUO
76633-500EA 579.4 USD
CA76633-498 76633-500 76633-498 CA76633-502 CA76633-500 76633-502
Anti-CD10 CALLA Mouse Monoclonal Antibody [clone: 56C6]
Antibodies
This antibody is specific to human CD10 antigen of 100 kDa, also known as common acute lymphocytic leukemia antigen (CALLA). CD10 antigen has been identified on the surface of early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue.