Anti-Chromogranin A Mouse Monoclonal Antibody [clone: LK2H10]
Supplier: Diagnostic BioSystems
PDM067 PDM067-RUO
76634-324EA 516.65 USD
76634-324 CA76634-324
Anti-Chromogranin A Mouse Monoclonal Antibody [clone: LK2H10]
Antibodies
Chromogranin A is a 439 amino acid protein present in secretory granules of a wide variety of endocrine cells and neurons. A positive staining is seen in the secretory granules of parathyroid, adrenal medulla, anterior pituitary gland and Langerhans islets of pancreas. This antibody stains a variety of neuroendocrine tumors.