Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: N10/2]
Supplier: Diagnostic BioSystems
MOB077-05-RUO MOB077-01-RUO MOB077 MOB077-01 MOB077-05 MOB077-RUO
76633-230EA 334.4 USD
CA76633-230 76633-230 CA76633-228 76633-228 CA76633-226 76633-226
Anti-Lambda Light Chain Mouse Monoclonal Antibody [clone: N10/2]
Antibodies
This antibody does not show any cross-reactivity with kappa light chain. It stains B cell follicles in human lymphoid tissues. The mantle zone shows a mosaic pattern of labeling while the germinal centers show a coarse meshwork pattern of staining.