Upright Industrial Microscopes, PA53MET
Supplier: Motic
1100104600861 1100104600881 1100104600891
76637-482EA 13234.13 USD
76637-482 76637-484 76637-486
Upright Industrial Microscopes, PA53MET
Microscopes
Motic new PA53 MET microscopes introduce a superior optical performance, implemented in an ergonomic hardware setup, and combined with automated functions for precise quantifications. The Motic PA53 MET series: ultimate solutions for industrial quality control and analysis of opaque and transparent materials.
  • Scalable Infinity-Corrected Optical System (CCIS®)
  • Trinocular erect image observation tube head (FN25)
  • Widefield (FN25) eyepiece 10×/25 mm with adjustable diopter
A flexible, high-performance optical system for a large array of inspection tasks.More Product Information