Motic AE31E LED Inverted Microscope
Supplier: Motic
1100100200841 1100100200862
76595-436EA 4744.44 USD
76595-436 76595-438
Motic AE31E LED Inverted Microscope
Microscopes
The AE31 Elite is a refined inverted microscope with an extended focus on practical aspects. In order to harmonize the optical setup of our CCIS© infinity concept, the AE31 Elite works with the newest generation of LWD plan achromatic lenses.
  • Siedentopf binocular tube 45° inclined [trinocular: (light split: ²⁰/₈₀)]
  • Side facing quintuple nosepiece
  • ELWD N.A. 0.30 (W.D. 72 mm) condenser
  • Phase slider Ph0, BF, Ph1
  • Phase centering telescope
Binocular or trinocular live cell microscope platform for advanced microbiology. The powerful LWD CCIS© phaseMore Product Information