PCR Tube Strips, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0785/B 4TI-0785/G 4TI-0785/M 4TI-0785/P 4TI-0785/R 4TI-0785/W/2D 4TI-0785/X 4TI-0785/X/2D 4TI-0785/XW 4TI-0786/B 4TI-0786/G 4TI-0786/M 4TI-0786/P 4TI-0786/R 4TI-0786/W/2D 4TI-0786/X 4TI-0786/X/2D 4TI-0786/XW
76629-026CS 161.41 USD
76629-026 76629-016 76629-062 76629-156 76629-164 76629-124 76629-068 76629-044 76629-056 76629-140 76629-112 76629-024 76629-070 76629-074 76629-130 76629-022 76629-120 76629-092
PCR Tube Strips, Azenta Life Sciences
Microplates PCR Plates
Strips of eight polypropylene PCR tubes held in a rigid polycarbonate frame for highest mechanical stability.
  • Two component design in an eight well strip format
  • Compatible with the majority of thermal cyclers
  • Available with either domed cap strips or with flat, optically clear cap strips
  • Available with off-the-shelf 2D code
By molding the frame portion in a more rigid polymer, the mechanical stability is greatly improved compared with traditional single piece products.
For color coding of experiments, we offer strips with clear wells in six different frame colors. Additionally, for optical assays such as qPCR, we supply strips with white wells in black frames. White wells increase the signal-to-noise ratio by maximizing reflection of light in fluorescent based assays.
PCR tubes and strips are available with cap strips (domed or optically flat) or without cap strips and are compatible with the majority of thermal cyclers. End tabs allow for easy handling and labelling of the strips and some products are also available with an off-the-shelf 2D code, offering a vast supply of unique code combinations.