PCR Plate Segments, Non-Skirted, Azenta Life Sciences
Supplier: Azenta Life Sciences
4TI-0750/32/Y 4TI-0750/32/P 4TI-0750/24/Y 4TI-0750/16/R 4TI-0750/8/R 4TI-0750/32/G 4TI-0750/32/B 4TI-0750/32 4TI-0750/16/G 4TI-0750/16/Y 4TI-0750/8/Y 4TI-0750/48 4TI-0750/24 4TI-0750/24/R 4TI-0750/32/R 4TI-0750/48/B 4TI-0750/48/G 4TI-0750/16/B 4TI-0750/24/G 4TI-0750/24/B 4TI-0750/8/G 4TI-0750/8/P 4TI-0750/16/P 4TI-0750/48/P 4TI-0750/8 4TI-0750/24/P 4TI-0750/48/Y 4TI-0750/48/R 4TI-0750/8/B 4TI-0750/16
76638-080CS 259.48 USD
76638-080 76637-972 76637-970 76637-992 76638-042 76638-240 76638-184 76638-044 76638-000 76638-088 76638-120 76638-142 76638-180 76638-116 76638-112 76638-014 76638-090 76637-984 76638-074 76638-052 76638-032 76638-192 76638-092 76638-194 76638-050 76638-028 76638-026 76638-146 76638-168 76637-990
PCR Plate Segments, Non-Skirted, Azenta Life Sciences
Microplates PCR Plates
0.2 ml wells, polypropylene, cut corner H12; working volume: <200 µl, total well capacity: 300 µl.
  • Subject to a strict QC procedure which includes a visual check and leak testing of every tube
  • Free from DNase, RNase, human genomic DNA and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Low profile 0.2 ml (200 µl) working volume, 0.3 ml (300 µl) total well capacity when used with sealing options
  • Maximum thermal conductivity for efficient heat transfer and precise thermal cycling
  • Raised well rims prevent cross contamination and facilitate effective sealing for reduced evaporation
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
  • Compatible with universal standard block thermal cyclers, sequencers and standard multichannel pipettes
These plates are produced by dividing the 96 well non-skirted PCR plate into smaller segments, a versatile solution for when a whole PCR plate may not be needed.
All of these PCR plates are molded from virgin polypropylene under ISO certified cleanroom conditions in a UK-based production facility, and as such comply with the same stringent requirements as the FrameStar® range.