Urine Cytology Collection Kit, StatLab
Supplier: StatLab
UCK-TP
76548-300EA 17.84 USD
76548-300
Urine Cytology Collection Kit, StatLab
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Convenient method for collection, preservation, and transport of urine cytology specimens. For urine cytological testing, kit contains prefilled fixative jar, sterile 120 mL urine collection jar, and a biohazard transport bag.
Packaging: Manufactured in the USA