Nail Prep™ Nail Softener, StatLab
Supplier: StatLab
NP-500
76548-176EA 141.66 USD
76548-176
Nail Prep™ Nail Softener, StatLab
Stains Microscopy and staining reagents
Try this exclusive formulation designed for pre-processing nail preparation. Fast, efficient, and ready to use, with two concentrations available. Use pre-processing. The solution softens brittle nail clippings. Nail Prep™ makes specimens easier to cut and process, and helps to keep tissue from washing off the slide.
Packaging: Manufactured in the USA